Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Synthware
™
gas washing bottle with fritted disc
Products
Product Category
bottles (6)
Manufacturer
Capacity (mL)
Height (mm)
Cap
Color
Vial Bottle Color
Package Size
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
SYNB300125C
Synthware
™
gas washing bottle with fritted disc,
125 mL, joint: ST/NS 45/50, Disc porosity C
Expand
View Pricing
SYNB300500C
Synthware
™
gas washing bottle with fritted disc,
500 mL, joint: ST/NS 45/50, Disc porosity C
Expand
View Pricing
SYNB310350C
Synthware
™
gas washing bottle with fritted disc,
joint: ST/NS 24/40, capacity 350 mL, Disc porosity C
Expand
View Pricing
SYNB300500X
Synthware
™
gas washing bottle with fritted disc,
500 mL, joint: ST/NS 45/50, Disc porosity XC
Expand
View Pricing
SYNB300250C
Synthware
™
gas washing bottle with fritted disc,
250 mL, joint: ST/NS 45/50, Disc porosity C
Expand
View Pricing
SYNB300125X
Synthware
™
gas washing bottle with fritted disc,
125 mL, joint: ST/NS 45/50, Disc porosity XC
Expand
View Pricing