Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Synthware
™
Erlenmeyer flask
Products
Product Category
flasks (18)
Manufacturer
Capacity (mL)
Cap
Color
Sterility
Filter Products
Showing 1-18 of 18
Compare
Product Number
Description
Pricing
SYNF664250
Synthware
™
Erlenmeyer flask,
capacity 250 mL, joint: ST/NS 24/40
Expand
View Pricing
SYNF664500
Synthware
™
Erlenmeyer flask,
capacity 500 mL, joint: ST/NS 24/40
Expand
View Pricing
SYNF662450
Synthware
™
Erlenmeyer flask,
capacity 50 mL, joint: ST/NS 24/40
Expand
View Pricing
SYNF501010
Synthware
™
Erlenmeyer flask,
capacity 10 mL, joint: ST/NS 14/10
Expand
View Pricing
SYNF664000
Synthware
™
Erlenmeyer flask,
capacity 1,000 mL, joint: ST/NS 24/40
Expand
View Pricing
SYNF661405
Synthware
™
Erlenmeyer flask,
capacity 5 mL, joint: ST/NS 14/20
Expand
View Pricing
SYNF664125
Synthware
™
Erlenmeyer flask,
capacity 125 mL, joint: ST/NS 24/40
Expand
View Pricing
SYNF668000
Synthware
™
Erlenmeyer flask,
capacity 1,000 mL, joint: ST/NS 29/42
Expand
View Pricing
SYNF661425
Synthware
™
Erlenmeyer flask,
capacity 25 mL, joint: ST/NS 14/20
Expand
View Pricing
SYNF661950
Synthware
™
Erlenmeyer flask,
capacity 50 mL, joint: ST/NS 19/22
Expand
View Pricing
SYNF669250
Synthware
™
Erlenmeyer flask,
capacity 250 mL, joint: ST/NS 29/42
Expand
View Pricing
SYNF662950
Synthware
™
Erlenmeyer flask,
capacity 50 mL, joint: ST/NS 29/42
Expand
View Pricing
SYNF669125
Synthware
™
Erlenmeyer flask,
capacity 125 mL, joint: ST/NS 19/22
Expand
View Pricing
SYNF501025
Synthware
™
Erlenmeyer flask,
capacity 25 mL, joint: ST/NS 14/10
Expand
View Pricing
SYNF669500
Synthware
™
Erlenmeyer flask,
capacity 500 mL, joint: ST/NS 29/42
Expand
View Pricing
SYNF661125
Synthware
™
Erlenmeyer flask,
capacity 125 mL, joint: ST/NS 14/20
Expand
View Pricing
SYNF661450
Synthware
™
Erlenmeyer flask,
capacity 50 mL, joint: ST/NS 14/20
Expand
View Pricing
SYNF661410
Synthware
™
Erlenmeyer flask,
joint: ST/NS 14/20, capacity 10 mL
Expand
View Pricing