Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Synthware
™
Crystallizing dish with spout
Products
Manufacturer
Diameter (mm)
Volume (ml)
Color
Package Size
Style
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
SYNP291200
Synthware
™
Crystallizing dish with spout,
clear borosilicate glass 3.3, volume 900 mL, diam. × H 150 mm × 75 mm
Expand
View Pricing
SYNP290170
Synthware
™
Crystallizing dish with spout,
clear borosilicate glass 3.3, volume 150 mL, diam. × H 80 mm × 40 mm
Expand
View Pricing
SYNP290700
Synthware
™
Crystallizing dish with spout,
clear borosilicate glass 3.3, volume 250 mL, diam. × H 100 mm × 50 mm
Expand
View Pricing