
SyntheChol® NS0 Supplement

Synonym(s):
Cholesterol solution
Empirical Formula (Hill Notation):
C27H46O
CAS Number:
Molecular Weight:
386.65
Products
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Product Line
Quality Segment
Sterilization Method
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing