Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Supel
™
Swift HLB DPX Tip
Synonym(s):
Supel
™
Swift HLB DPX SPE
Products
Brand
Supelco (7)
Volume (ml)
Volume (µL)
Compatibility
Quality Segment
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
52989-U
Supel
™
Swift HLB DPX Tip,
Universal format, Tip volume 1 mL, box of 96 tips, bed weight 5 mg
Expand
View Pricing
52992-U
Supel
™
Swift HLB DPX Tip,
Hamilton, Tip volume 1 mL, box of 96 tips, bed weight 10 mg
Expand
View Pricing
52999-U
Supel
™
Swift HLB DPX Tip,
Universal format, Tip volume 1 mL, box of 96 tips, bed weight 20 mg
Expand
View Pricing
53001-U
Supel
™
Swift HLB DPX Tip,
Hamilton, Tip volume 300 μL, box of 96 tips, bed weight 3 mg
Expand
View Pricing
52995-U
Supel
™
Swift HLB DPX Tip,
Universal format, Tip volume 1 mL, box of 96 tips, bed weight 10 mg
Expand
View Pricing
52984-U
Supel
™
Swift HLB DPX Tip,
Hamilton, Tip volume 1 mL, box of 96 tips, bed weight 5 mg
Expand
View Pricing
52997-U
Supel
™
Swift HLB DPX Tip,
Hamilton, Tip volume 1 mL, box of 96 tips, bed weight 20 mg
Expand
View Pricing