Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Supel
™
-Inert Multi-Layer Foil
Synonym(s):
Supel
™
-Inert Multi-Layer Foil
Products
Brand
Supelco (8)
Product Line
Agency
Length (in)
Application
Package Size
Technique
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
30228-U
Supel
™
-Inert Multi-Layer Foil,
maximum volume 5 L, Screw Cap Valve (SCV), pkg of 10 bags
Expand
View Pricing
30226-U
Supel
™
-Inert Multi-Layer Foil,
maximum volume 1 L, Screw Cap Valve (SCV), pkg of 10 bags
Expand
View Pricing
30227-U
Supel
™
-Inert Multi-Layer Foil,
maximum volume 2 L, Screw Cap Valve (SCV), pkg of 10 bags
Expand
View Pricing
30279-U
Supel
™
-Inert Multi-Layer Foil,
maximum volume 0.6 L, Screw Cap Valve (SCV), pkg of 10 bags
Expand
View Pricing
30229-U
Supel
™
-Inert Multi-Layer Foil,
maximum volume 10 L, Screw Cap Valve (SCV), pkg of 10 bags
Expand
View Pricing
30237-U
Supel
™
-Inert Multi-Layer Foil,
maximum volume 1 L, Push/Pull Lock Valve (PLV), pkg of 10 bags
Expand
View Pricing
30281-U
Supel
™
-Inert Multi-Layer Foil,
maximum volume 0.6 L, Push/Pull Lock Valve (PLV), pkg of 10 bags
Expand
View Pricing
30238-U
Supel
™
-Inert Multi-Layer Foil,
maximum volume 2 L, Push/Pull Lock Valve (PLV), pkg of 10 bags
Expand
View Pricing
30240-U
Supel
™
-Inert Multi-Layer Foil,
maximum volume 10 L, Push/Pull Lock Valve (PLV), pkg of 10 bags
Expand
View Pricing