
Sulfosuccinic acid solution

Synonym(s):
2-Sulfobutanedioic acid
Linear Formula:
HOOCCH2CH(SO3H)COOH
CAS Number:
Molecular Weight:
198.15
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing