Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sulforhodamine B sodium salt
Empirical Formula (Hill Notation):
C
27
H
29
N
2
NaO
7
S
2
CAS Number:
3520-42-1
Molecular Weight:
580.65
EC Number:
222-529-8
Products
Product Category
stains (1)
Biological Source
Formula Weight
Physical Form
Product Line
Chemical Composition
Application
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
S1402
Sulforhodamine B sodium salt,
powder, BioReagent, suitable for cell culture
Expand
View Pricing
S9012
Sulforhodamine B sodium salt,
Powder
Expand
View Pricing