
Succinylmonocholine chloride

Synonym(s):
2-(3-Carboxy-1-oxopropoxy)-N,N,N-trimethylethanaminium chloride, Succinylmonocholine chloride
Empirical Formula (Hill Notation):
C9H18ClNO4
CAS Number:
Molecular Weight:
239.70
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2