Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Strontium carbonate
Linear Formula:
SrCO
3
CAS Number:
1633-05-2
Molecular Weight:
147.63
Products
Product Category
inorganic bases (3)
Brand
Sigma-Aldrich (3)
Formula Weight
Manufacturer
Physical Form
Purity
Format
Quality Segment
Special Grade
certified reference material (1)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
472018
Strontium carbonate,
≥99.9% trace metals basis
Expand
View Pricing
289833
Strontium carbonate,
≥98%
Expand
View Pricing
204455
Strontium carbonate,
99.995% trace metals basis
Expand
View Pricing
NIST987
Strontium carbonate,
NIST
®
SRM
®
987, isotopic standard
Expand
View Pricing
1.07861
Strontium carbonate,
Technipur
®
Expand
View Pricing