Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Starch from corn
Synonym(s):
Corn starch, Starch from maize
Linear Formula:
(C
6
H
10
O
5
)
n
CAS Number:
9005-25-8
EC Number:
232-679-6
Products
Product Category
polysaccharides (4)
carbohydrates (3)
Brand
Sigma-Aldrich (1)
Supelco (2)
Biological Source
Color
Feature
Physical Form
Format
Analyte Suitability
Quality Segment
Application
Reference Material Type
Special Grade
Practical (1)
analytical standard (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
S4126
Starch from corn,
Expand
View Pricing
S4180
Starch from corn,
practical grade
Expand
View Pricing
S9679
Starch from corn,
Unmodified waxy corn starch of essentially pure amylopectin; contains only trace amounts of amylose.
Expand
View Pricing
S5296
Starch from corn,
analytical standard, analytical standard for Starch Assay Kits SA-20 and STA-20
Expand
View Pricing
S2388
Starch from corn,
for use with Total Dietary Fiber Control Kit, TDF-C10
Expand
View Pricing