Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Stainless steel - AISI 347
Products
Manufacturer
Physical Form
Length (m)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
GF31952088
Stainless steel - AISI 347,
wire, Fe/Cr18%/Ni 10%/Nb, 0.5 mm diameter, length 10 m, temper annealed
Expand
View Pricing
GF41529067
Stainless steel - AISI 347,
foil, Fe/Cr18%/Ni 10%/Nb, thickness 0.05 mm, size 25 × 25 mm, temper annealed
Expand
View Pricing
GF94677733
Stainless steel - AISI 347,
powder, 45 max. part. size (micron), Fe/Cr18%/Ni10%/Nb%, weight 50 g, condition atomized
Expand
View Pricing
GF91133298
Stainless steel - AISI 347,
foil, thickness 0.05 mm, Fe/Cr18%/Ni10%/Nb%, size 50 × 50 mm, temper annealed
Expand
View Pricing