Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Stainless Steel - AISI 316L alloy, FeCr18Ni10Mo3
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Length (mm)
Filter Products
Showing 1-20 of 201
Page 1 of 11
Page 1 of 11
Compare
Product Number
Description
Pricing
GF68846552
Stainless Steel - AISI 316L alloy, FeCr18Ni10Mo3,
powder, 3micron mean particle size, atomized, 50g
Expand
View Pricing
GF40713612
Stainless Steel - AISI 316L alloy, FeCr18Ni10Mo3,
500mm tube, 4.2mm outside diameter, 0.2mm wall thickness, 3.8mm inside diameter, annealed
Expand
View Pricing
GF43365001
Stainless Steel - AISI 316L alloy, FeCr18Ni10Mo3,
powder, 3micron mean particle size, atomized, 100g
Expand
View Pricing
GF33328582
Stainless Steel - AISI 316L alloy, FeCr18Ni10Mo3,
foil, 100 x 100mm, 0.1mm thickness, annealed
Expand
View Pricing
GF02730484
Stainless Steel - AISI 316L alloy, FeCr18Ni10Mo3,
foil, 6mm disks, 0.2mm thickness, annealed
Expand
View Pricing
GF77510707
Stainless Steel - AISI 316L alloy, FeCr18Ni10Mo3,
powder, 3micron mean particle size, atomized, 200g
Expand
View Pricing
GF10003239
Stainless Steel - AISI 316L alloy, FeCr18Ni10Mo3,
foil, 50 x 50mm, 0.5mm thickness, annealed, polished on both sides
Expand
View Pricing
GF52037029
Stainless Steel - AISI 316L alloy, FeCr18Ni10Mo3,
1000mm rod, 25.4mm diameter
Expand
View Pricing
GF96939253
Stainless Steel - AISI 316L alloy, FeCr18Ni10Mo3,
foil, 100 x 100mm, 0.5mm thickness, annealed, polished on both sides
Expand
View Pricing
GF28668808
Stainless Steel - AISI 316L alloy, FeCr18Ni10Mo3,
foil, 6mm disks, 0.1mm thickness, annealed
Expand
View Pricing
GF05917978
Stainless Steel - AISI 316L alloy, FeCr18Ni10Mo3,
foil, 100 x 100mm, 0.9mm thickness, annealed, mirror polished on both sides
Expand
View Pricing
GF13826437
Stainless Steel - AISI 316L alloy, FeCr18Ni10Mo3,
1000mm wire, 0.25mm diameter, annealed, straight wire
Expand
View Pricing
GF50260328
Stainless Steel - AISI 316L alloy, FeCr18Ni10Mo3,
100m wire, 0.5mm diameter, as drawn
Expand
View Pricing
GF56067497
Stainless Steel - AISI 316L alloy, FeCr18Ni10Mo3,
foil, 25mm disks, 0.38mm thickness, annealed
Expand
View Pricing
GF05332698
Stainless Steel - AISI 316L alloy, FeCr18Ni10Mo3,
1000mm tube, 6.0mm outside diameter, 1.0mm wall thickness, 4mm inside diameter, annealed
Expand
View Pricing
GF63756153
Stainless Steel - AISI 316L alloy, FeCr18Ni10Mo3,
1000mm wire, 0.5mm diameter, annealed, straight wire
Expand
View Pricing
GF61395745
Stainless Steel - AISI 316L alloy, FeCr18Ni10Mo3,
50m wire, 0.7mm diameter, as drawn
Expand
View Pricing
GF16698033
Stainless Steel - AISI 316L alloy, FeCr18Ni10Mo3,
100m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF43061352
Stainless Steel - AISI 316L alloy, FeCr18Ni10Mo3,
foil, 50 x 50mm, 0.9mm thickness, annealed, mirror polished on both sides
Expand
View Pricing
GF18195082
Stainless Steel - AISI 316L alloy, FeCr18Ni10Mo3,
foil, 0.5m coil, 12mm coil width, 0.2mm thickness, annealed
Expand
View Pricing
Page 1 of 11
Page 1 of 11