Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-20 of 201
Page 1 of 11
Page 1 of 11
Compare
Product Number
Description
Pricing
GF36611805
Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3,
mesh, 600 x 600mm, 0.103mm nominal aperture, 0.066mm wire diameter, 150 x 150 wires/inch, 0.37 open area, plain weave mesh type
Expand
View Pricing
GF08787842
Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3,
mesh, 100 x 100mm, 0.38mm nominal aperture, 0.25mm wire diameter, 40 x 40 wires/inch, 0.37 open area, plain weave mesh type
Expand
View Pricing
GF70782240
Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3,
mesh, 300 x 300mm, 0.103mm nominal aperture, 0.066mm wire diameter, 150 x 150 wires/inch, 0.37 open area, plain weave mesh type
Expand
View Pricing
GF68848521
Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3,
mesh, 150 x 150mm, 0.103mm nominal aperture, 0.066mm wire diameter, 150 x 150 wires/inch, 0.37 open area, plain weave mesh type
Expand
View Pricing
GF87616851
Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3,
powder, 45micron max. particle size, 100g
Expand
View Pricing
GF98480683
Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3,
mesh, 150 x 150mm, 0.38mm nominal aperture, 0.25mm wire diameter, 40 x 40 wires/inch, 0.37 open area, plain weave mesh type
Expand
View Pricing
GF82981996
Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3,
foil, 6mm disks, 0.0125mm thickness, hard
Expand
View Pricing
GF26340689
Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3,
powder, 150micron max. particle size, 200g
Expand
View Pricing
GF60776668
Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3,
mesh, 600 x 600mm, 0.38mm nominal aperture, 0.25mm wire diameter, 40 x 40 wires/inch, 0.37 open area, plain weave mesh type
Expand
View Pricing
GF10086218
Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3,
mesh, 300 x 300mm, 0.38mm nominal aperture, 0.25mm wire diameter, 40 x 40 wires/inch, 0.37 open area, plain weave mesh type
Expand
View Pricing
GF59805603
Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3,
500mm tube, 8.0mm outside diameter, 1.0mm wall thickness, 6mm inside diameter, annealed
Expand
View Pricing
GF49810721
Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3,
powder, 150micron max. particle size, 100g
Expand
View Pricing
GF43613726
Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3,
sphere, 0.5mm diameter, 2.50μm sphericity, precision sphere, grade 100, 200pcs
Expand
View Pricing
GF64704685
Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3,
sphere, 1.0mm diameter, 2.50μm sphericity, precision sphere, grade 100, 100pcs
Expand
View Pricing
GF22429166
Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3,
foil, 50 x 50mm, 0.1mm thickness, annealed
Expand
View Pricing
GF37437307
Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3,
foil, 0.5m coil, 0.025mm thickness, annealed
Expand
View Pricing
GF60980081
Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3,
powder, 45micron max. particle size, 1000g
Expand
View Pricing
GF55309787
Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3,
20m insulated wire, 0.075mm conductor diameter, 0.018mm insulation thickness, ptfe (polytetrafluoroethylene) insulation
Expand
View Pricing
GF46625193
Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3,
foil, 25 x 25mm, 0.1mm thickness, hard
Expand
View Pricing
GF74132333
Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3,
sphere, 2.0mm diameter, 2.50μm sphericity, precision sphere, grade 100, 100pcs
Expand
View Pricing
Page 1 of 11
Page 1 of 11