Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Stainless Steel - AISI 304 alloy, FeCr18Ni10
Products
Manufacturer
Physical Form
ID (mm)
Length (mm)
Filter Products
Showing 1-20 of 201
Page 1 of 11
Page 1 of 11
Compare
Product Number
Description
Pricing
GF53057099
Stainless Steel - AISI 304 alloy, FeCr18Ni10,
mesh, 100 x 100mm, 0.38mm nominal aperture, 0.75mm thickness, 0.25mm wire diameter, 40 x 40 wires/inch, 0.37 open area, plain weave mesh
Expand
View Pricing
GF95455402
Stainless Steel - AISI 304 alloy, FeCr18Ni10,
100m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF02863841
Stainless Steel - AISI 304 alloy, FeCr18Ni10,
powder, 45micron max. particle size, 100g
Expand
View Pricing
GF08394497
Stainless Steel - AISI 304 alloy, FeCr18Ni10,
powder, 45micron max. particle size, 2000g
Expand
View Pricing
GF99413499
Stainless Steel - AISI 304 alloy, FeCr18Ni10,
200mm tube, 0.8mm outside diameter, 0.15mm wall thickness, 0.5mm inside diameter, annealed
Expand
View Pricing
GF17063490
Stainless Steel - AISI 304 alloy, FeCr18Ni10,
mesh, 300 x 300mm, 0.38mm nominal aperture, 0.75mm thickness, 0.25mm wire diameter, 40 x 40 wires/inch, 0.37 open area, plain weave mesh
Expand
View Pricing
GF52867154
Stainless Steel - AISI 304 alloy, FeCr18Ni10,
powder, 45micron max. particle size, 1000g
Expand
View Pricing
GF12578799
Stainless Steel - AISI 304 alloy, FeCr18Ni10,
sphere, 0.5mm diameter, 2.50μm sphericity, precision sphere, grade 100, 100pcs
Expand
View Pricing
GF32136500
Stainless Steel - AISI 304 alloy, FeCr18Ni10,
mesh, 150 x 150mm, 0.38mm nominal aperture, 0.75mm thickness, 0.25mm wire diameter, 40 x 40 wires/inch, 0.37 open area, plain weave mesh
Expand
View Pricing
GF17132239
Stainless Steel - AISI 304 alloy, FeCr18Ni10,
mesh, 900 x 900mm, 0.38mm nominal aperture, 0.75mm thickness, 0.25mm wire diameter, 40 x 40 wires/inch, 0.37 open area, plain weave mesh
Expand
View Pricing
GF37396424
Stainless Steel - AISI 304 alloy, FeCr18Ni10,
100mm tube, 1.1mm outside diameter, 0.2mm wall thickness, 0.7mm inside diameter, hard
Expand
View Pricing
GF26509989
Stainless Steel - AISI 304 alloy, FeCr18Ni10,
500mm tube, 10.0mm outside diameter, 2.0mm wall thickness, 6mm inside diameter, annealed
Expand
View Pricing
GF52802603
Stainless Steel - AISI 304 alloy, FeCr18Ni10,
1000m wire, 0.05mm diameter, annealed
Expand
View Pricing
GF93979298
Stainless Steel - AISI 304 alloy, FeCr18Ni10,
foil, 1m coil, 0.2mm thickness, annealed
Expand
View Pricing
GF36600599
Stainless Steel - AISI 304 alloy, FeCr18Ni10,
foil, 100 x 100mm, 0.025mm thickness, hard, light tight (lt)
Expand
View Pricing
GF57621999
Stainless Steel - AISI 304 alloy, FeCr18Ni10,
foil, 100 x 100mm, 0.015mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF26169644
Stainless Steel - AISI 304 alloy, FeCr18Ni10,
foil, 300 x 300mm, 0.015mm thickness, as rolled, not light tested (nlt)
Expand
View Pricing
GF49300051
Stainless Steel - AISI 304 alloy, FeCr18Ni10,
1000mm wire, 1.0mm diameter, as drawn, straight wire
Expand
View Pricing
GF29417148
Stainless Steel - AISI 304 alloy, FeCr18Ni10,
500mm tube, 2.05mm outside diameter, 0.25mm wall thickness, 1.55mm inside diameter, hard
Expand
View Pricing
GF37298931
Stainless Steel - AISI 304 alloy, FeCr18Ni10,
100mm tube, 0.5mm outside diameter, 0.11mm wall thickness, 0.28mm inside diameter, hard
Expand
View Pricing
Page 1 of 11
Page 1 of 11