Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Stainless Steel - AISI 302 alloy, FeCr18Ni8
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-20 of 138
Page 1 of 7
Page 1 of 7
Compare
Product Number
Description
Pricing
GF30258491
Stainless Steel - AISI 302 alloy, FeCr18Ni8,
50m wire, 0.05mm diameter, hard
Expand
View Pricing
GF69014628
Stainless Steel - AISI 302 alloy, FeCr18Ni8,
25m wire, 0.25mm diameter, hard
Expand
View Pricing
GF83254181
Stainless Steel - AISI 302 alloy, FeCr18Ni8,
100m wire, 0.025mm diameter, annealed
Expand
View Pricing
GF33959403
Stainless Steel - AISI 302 alloy, FeCr18Ni8,
5m wire, 0.5mm diameter, hard
Expand
View Pricing
GF32317321
Stainless Steel - AISI 302 alloy, FeCr18Ni8,
5m wire, 0.01mm diameter, as drawn
Expand
View Pricing
GF69778300
Stainless Steel - AISI 302 alloy, FeCr18Ni8,
100m wire, 0.5mm diameter, hard
Expand
View Pricing
GF15758220
Stainless Steel - AISI 302 alloy, FeCr18Ni8,
50m wire, 0.5mm diameter, hard
Expand
View Pricing
GF33459510
Stainless Steel - AISI 302 alloy, FeCr18Ni8,
foil, 100 x 100mm, 0.2mm thickness, hard
Expand
View Pricing
GF90729838
Stainless Steel - AISI 302 alloy, FeCr18Ni8,
100m wire, 0.1mm diameter, annealed
Expand
View Pricing
GF38989591
Stainless Steel - AISI 302 alloy, FeCr18Ni8,
25m wire, 0.1mm diameter, annealed
Expand
View Pricing
GF83317349
Stainless Steel - AISI 302 alloy, FeCr18Ni8,
foil, 25 x 25mm, 0.2mm thickness, hard
Expand
View Pricing
GF44926061
Stainless Steel - AISI 302 alloy, FeCr18Ni8,
10m wire, 0.5mm diameter, hard
Expand
View Pricing
GF23843011
Stainless Steel - AISI 302 alloy, FeCr18Ni8,
25m wire, 0.125mm diameter, hard
Expand
View Pricing
GF35019345
Stainless Steel - AISI 302 alloy, FeCr18Ni8,
foil, 25 x 25mm, 0.15mm thickness, hard
Expand
View Pricing
GF49089392
Stainless Steel - AISI 302 alloy, FeCr18Ni8,
5m insulated wire, 0.1mm conductor diameter, 0.01mm insulation thickness, epoxy insulation
Expand
View Pricing
GF22665078
Stainless Steel - AISI 302 alloy, FeCr18Ni8,
200mm wire, 0.1mm diameter, hard, straight wire
Expand
View Pricing
GF55601298
Stainless Steel - AISI 302 alloy, FeCr18Ni8,
100m wire, 0.1mm diameter, hard
Expand
View Pricing
GF69842065
Stainless Steel - AISI 302 alloy, FeCr18Ni8,
25m wire, 0.05mm diameter, hard
Expand
View Pricing
GF34199376
Stainless Steel - AISI 302 alloy, FeCr18Ni8,
foil, 2m coil, 0.2mm thickness, hard
Expand
View Pricing
GF26246347
Stainless Steel - AISI 302 alloy, FeCr18Ni8,
foil, 25mm disks, 0.1mm thickness, hard
Expand
View Pricing
Page 1 of 7
Page 1 of 7