Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
SPME fiber assembly Polydimethylsiloxane (PDMS)
Products
Brand
Supelco (6)
Sustainability
Feature
Length (cm)
Quality Segment
Application
Color
d
f
(µm)
Gauge
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
57301
SPME fiber assembly Polydimethylsiloxane (PDMS),
d
f
100 μm(PDMS, for use with autosampler, needle size 24 ga
Expand
View Pricing
57308
SPME fiber assembly Polydimethylsiloxane (PDMS),
d
f
30 μm(PDMS, needle size 24 ga, for use with manual holder
Expand
View Pricing
57302
SPME fiber assembly Polydimethylsiloxane (PDMS),
d
f
7 μm(PDMS, needle size 24 ga, for use with manual holder
Expand
View Pricing
57309
SPME fiber assembly Polydimethylsiloxane (PDMS),
d
f
30 μm(PDMS, needle size 24 ga, for use with autosampler
Expand
View Pricing
57303
SPME fiber assembly Polydimethylsiloxane (PDMS),
d
f
7 μm(PDMS, needle size 24 ga, for use with autosampler
Expand
View Pricing
57928-U
SPME fiber assembly Polydimethylsiloxane (PDMS),
metal alloy fiber, d
f
100 μm(PDMS, needle size 23 ga, for use with autosampler
Expand
View Pricing