Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Spinbar
®
magnetic stir bar
Products
Product Category
stir bars (63)
Brand
Sigma-Aldrich (37)
Color
Manufacturer
Diameter (mm)
Length (mm)
Package Size
Filter Products
Showing 1-20 of 63
Page 1 of 4
Page 1 of 4
Compare
Product Number
Description
Pricing
Z105066
Spinbar
®
magnetic stir bar,
PTFE-coated, octagonal, size 3 in. × 1/2 in., white
Expand
View Pricing
Z115304
Spinbar
®
magnetic stir bar,
egg-shaped PTFE-coated, size 3/4 in. × 3/8 in., white
Expand
View Pricing
Z126942
Spinbar
®
magnetic stir bar,
PTFE-coated, octagonal, size 1/2 in. × 1/8 in., white
Expand
View Pricing
Z266299
Spinbar
®
magnetic stir bar,
PTFE-coated, polygon, diam. 6 mm, L 15 mm, white
Expand
View Pricing
Z127027
Spinbar
®
magnetic stir bar,
PTFE-coated, octagonal, size 2 1/2 in. × 5/16 in., white
Expand
View Pricing
Z105643
Spinbar
®
magnetic stir bar,
egg-shaped PTFE-coated, size 1 in. × 1/2 in., white
Expand
View Pricing
Z266329
Spinbar
®
magnetic stir bar,
PTFE-coated, polygon, diam. 8 mm, L 30 mm, white
Expand
View Pricing
Z105678
Spinbar
®
magnetic stir bar,
egg-shaped PTFE-coated, size 1 1/2 in. × 5/8 in., white
Expand
View Pricing
Z105651
Spinbar
®
magnetic stir bar,
egg-shaped PTFE-coated, size 1 1/4 in. × 5/8 in., white
Expand
View Pricing
Z126950
Spinbar
®
magnetic stir bar,
PTFE-coated, octagonal, size 1/2 in. × 5/16 in., white
Expand
View Pricing
Z127019
Spinbar
®
magnetic stir bar,
PTFE-coated, octagonal, size 2 in. × 5/16 in., white
Expand
View Pricing
Z175412
Spinbar
®
magnetic stir bar,
egg-shaped PTFE-coated, size 3/8 in. × 3/16 in., white
Expand
View Pricing
Z266337
Spinbar
®
magnetic stir bar,
PTFE-coated, polygon, diam. 8 mm, L 40 mm, white
Expand
View Pricing
Z284483
Spinbar
®
magnetic stir bar,
round PTFE-coated, size 2 in. × 5/16 in., white
Expand
View Pricing
Z115339
Spinbar
®
magnetic stir bar,
egg-shaped PTFE-coated, size 3.0 in. × 3/4 in., white
Expand
View Pricing
Z127078
Spinbar
®
magnetic stir bar,
PTFE-coated, octagonal, size 1 in. × 3/8 in., white
Expand
View Pricing
Z266388
Spinbar
®
magnetic stir bar,
PTFE-coated, polygon, diam. 27 mm, L 108 mm, white
Expand
View Pricing
Z127116
Spinbar
®
magnetic stir bar,
PTFE-coated, octagonal, size 1 1/2 in. × 1/2 in., white
Expand
View Pricing
Z104442
Spinbar
®
magnetic stir bar,
PTFE-coated, octagonal, size 1 1/2 in. × 5/16 in., white
Expand
View Pricing
Z104450
Spinbar
®
magnetic stir bar,
PTFE-coated, octagonal, size 2 in. × 3/8 in., white
Expand
View Pricing
Page 1 of 4
Page 1 of 4