Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
SP Bel-Art
®
utility bags
Products
Product Category
bags (21)
Manufacturer
Color
Package Size
Filter Products
Showing 1-20 of 21
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
BAH131780610
SP Bel-Art
®
utility bags,
width × H × thickness 6 in. (152 mm) × 10 in. (254 mm) × 4 mil (0.1 mm), clear, pack of 100 ea
Expand
View Pricing
BAH131780812
SP Bel-Art
®
utility bags,
width × H × thickness 8 in. (203 mm) × 12 in. (305 mm) × 4 in. (0.1 mm), clear, pack of 100 ea
Expand
View Pricing
BAH131760612
SP Bel-Art
®
utility bags,
width × H × thickness 6 in. (152 mm) × 12 in. (305 mm) × 1.5 mil (0.038 mm), clear, pack of 100 ea
Expand
View Pricing
BAH131780046
SP Bel-Art
®
utility bags,
width × H × thickness 4 in. (102 mm) × 6 in. (152 mm) × 4 mil (0.1 mm), clear, pack of 100 ea
Expand
View Pricing
BAH131780612
SP Bel-Art
®
utility bags,
width × H × thickness 6 in. (152 mm) × 12 in. (305 mm) × 4 mil (0.1 mm), clear, pack of 100 ea
Expand
View Pricing
BAH131780810
SP Bel-Art
®
utility bags,
width × H × thickness 8 mm (203) × 10.5 in. (267 mm) × 4 mil (0.1 mm), clear, pack of 100 ea
Expand
View Pricing
BAH131780068
SP Bel-Art
®
utility bags,
width × H × thickness 6 in. (152 mm) × 8.5 in. (216 mm) × 4 mil (0.1 mm), clear, pack of 100 ea
Expand
View Pricing
BAH131781218
SP Bel-Art
®
utility bags,
width × H × thickness 12 in. (305 mm) × 18 in. (457 mm) × 4 mil (0.1 mm), clear, pack of 100 ea
Expand
View Pricing
BAH131781220
SP Bel-Art
®
utility bags,
width × H × thickness 12 in. (305 mm) × 20 in. (508 mm) × 4 mil (0.1 mm), clear, pack of 100 ea
Expand
View Pricing
BAH131760812
SP Bel-Art
®
utility bags,
width × H × thickness 8 in. (203 mm) × 12 in. (305 mm) × 1.5 mil (0.038 mm), clear, pack of 100 ea
Expand
View Pricing
BAH131781016
SP Bel-Art
®
utility bags,
width × H × thickness 10 in. (254 mm) × 16 in. (406 mm) × 4 mil (0.1 mm), clear, pack of 100 ea
Expand
View Pricing
BAH131760818
SP Bel-Art
®
utility bags,
width × H × thickness 8 in. (203 mm) × 18 in. (457 mm) × 1.5 mil (0.038 mm), clear, pack of 100 ea
Expand
View Pricing
BAH131781018
SP Bel-Art
®
utility bags,
width × H × thickness 10 in. (254 mm) × 18 in. (457 mm) × 4 mil (0.1 mm), clear, pack of 100 ea
Expand
View Pricing
BAH131761218
SP Bel-Art
®
utility bags,
width × H × thickness 12 in. (305 mm) × 18 in. (457 mm) × 1.5 mil (0.038 mm), clear, pack of 100 ea
Expand
View Pricing
BAH131761014
SP Bel-Art
®
utility bags,
width × H × thickness 10 in. (254 mm) × 14 in. (356 mm) × 1.5 mil (0.038 mm), clear, pack of 100 ea
Expand
View Pricing
BAH131781012
SP Bel-Art
®
utility bags,
width × H × thickness 10 in. (254 mm) × 12 in. (305 mm) × 4 mil (0.1 mm), clear, pack of 100 ea
Expand
View Pricing
BAH131780816
SP Bel-Art
®
utility bags,
width × H × thickness 8 in. (203 mm) × 16 in. (406 mm) × 4 mil (0.1 mm), clear, pack of 100 ea
Expand
View Pricing
BAH131760816
SP Bel-Art
®
utility bags,
width × H × thickness 8 in. (203 mm) × 16 in. (406 mm) × 1.5 mil (0.038 mm), clear, pack of 100 ea
Expand
View Pricing
BAH131761012
SP Bel-Art
®
utility bags,
width × H × thickness 10 in. (254 mm) × 12 in. (305 mm) × 1.5 mil (0.038 mm), clear, pack of 100 ea
Expand
View Pricing
BAH131761016
SP Bel-Art
®
utility bags,
width × H × thickness 10 mm (254 in.) × 16 in. (406 mm) × 1.5 mil (0.038 mm), clear, pack of 100 ea
Expand
View Pricing
Page 1 of 2
Page 1 of 2