Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
SP Bel-Art
®
Straight Tubing Connectors
Products
Manufacturer
Package Size
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
BAH195620000
SP Bel-Art
®
Straight Tubing Connectors,
tubing I.D. 0.5-0.5 in. (6-12.7 mm)
Expand
View Pricing
BAH195080000
SP Bel-Art
®
Straight Tubing Connectors,
tubing I.D. 0.25 in. (6 mm)
Expand
View Pricing
BAH195070000
SP Bel-Art
®
Straight Tubing Connectors,
tubing I.D. 0.1875 in. (4.7 mm)
Expand
View Pricing
BAH195630000
SP Bel-Art
®
Straight Tubing Connectors,
tubing I.D. 0.5 in. (9.5-12.7 mm)
Expand
View Pricing
BAH195590000
SP Bel-Art
®
Straight Tubing Connectors,
tubing I.D. 0.5-0.5 in. (4.7-12.7 mm)
Expand
View Pricing
BAH195090000
SP Bel-Art
®
Straight Tubing Connectors,
tubing I.D. 0.375 in. (9.5 mm)
Expand
View Pricing
BAH195030000
SP Bel-Art
®
Straight Tubing Connectors,
tubing I.D. 0.125 in. (3.2 mm)
Expand
View Pricing
BAH195610000
SP Bel-Art
®
Straight Tubing Connectors,
tubing I.D. 0.375-0.375 in. (6-9.5 mm)
Expand
View Pricing
BAH195100000
SP Bel-Art
®
Straight Tubing Connectors,
tubing I.D. 0.5 in. (12.7 mm)
Expand
View Pricing
BAH195580000
SP Bel-Art
®
Straight Tubing Connectors,
tubing I.D. 0.25 in. (4.7-6 mm)
Expand
View Pricing