Skip to Content
Products
Cart
0
AF
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Back
SP Bel-Art
®
Sterileware
™
Teaspoon Style Sampling Spoon
Products
Product Category
spatulas/scoops (2)
Manufacturer
Height (mm)
Length (cm)
Length (in)
Length (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
BAF369410010
SP Bel-Art
®
Sterileware
™
Teaspoon Style Sampling Spoon,
white polystyrene, nominal capacity 10 mL (0.3 oz), handle L 13.7 cm (5.375 in.), sterile, pack of 100 ea (individually wrapped)
Expand
View Pricing
BAF369410003
SP Bel-Art
®
Sterileware
™
Teaspoon Style Sampling Spoon,
white polystyrene, nominal capacity 3 mL (0.1 oz), handle L 13.7 cm (5.375 in.), sterile, pack of 100 ea (individually wrapped)
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
English - EN
Learn More