Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
SP Bel-Art
®
Sterileware
™
Sampling Scoop with Handle
Products
Product Category
spatulas/scoops (14)
Manufacturer
Height (mm)
Length (cm)
Length (in)
Length (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
BAH369020000
SP Bel-Art
®
Sterileware
™
Sampling Scoop with Handle,
white polystyrene, nominal capacity 60 mL (2 oz), sterile, pkg of 100 ea (individually wrapped)
Expand
View Pricing
BAH369060000
SP Bel-Art
®
Sterileware
™
Sampling Scoop with Handle,
white polystyrene, nominal capacity 250 mL (8 oz), sterile, pack of 100 ea (individually wrapped)
Expand
View Pricing
BAH369020010
SP Bel-Art
®
Sterileware
™
Sampling Scoop with Handle,
white polystyrene, nominal capacity 60 mL (2 oz), sterile, pack of 10 ea (individually wrapped)
Expand
View Pricing
BAH369022002
SP Bel-Art
®
Sterileware
™
Sampling Scoop with Handle,
red polystyrene, nominal capacity 60 mL (2 oz), sterile, pack of 100 ea (individually wrapped)
Expand
View Pricing
BAH369060010
SP Bel-Art
®
Sterileware
™
Sampling Scoop with Handle,
white polystyrene, nominal capacity 250 mL (8 oz), sterile, pack of 10 ea (individually wrapped)
Expand
View Pricing
BAH369040000
SP Bel-Art
®
Sterileware
™
Sampling Scoop with Handle,
white polystyrene, nominal capacity 125 mL (4 oz), sterile, pack of 100 ea (individually wrapped)
Expand
View Pricing
BAH369021010
SP Bel-Art
®
Sterileware
™
Sampling Scoop with Handle,
white polystyrene, nominal capacity 60 mL, sterile, pack of 10 ea (double bagged)
Expand
View Pricing
BAH369062010
SP Bel-Art
®
Sterileware
™
Sampling Scoop with Handle,
red polystyrene, nominal capacity 250 mL (8 oz), sterile, pack of 100 ea (individually wrapped)
Expand
View Pricing
BAH369040010
SP Bel-Art
®
Sterileware
™
Sampling Scoop with Handle,
white polystyrene, nominal capacity 125 mL (4 oz), sterile, pack of 10 ea (individually wrapped)
Expand
View Pricing
BAH369022010
SP Bel-Art
®
Sterileware
™
Sampling Scoop with Handle,
red polystyrene, nominal capacity 60 mL (2 oz), sterile, pack of 10 ea (individually wrapped)
Expand
View Pricing
BAH369061010
SP Bel-Art
®
Sterileware
™
Sampling Scoop with Handle,
white polystyrene, nominal capacity 250 mL (8 oz), sterile, pack of 10 ea (double bagged)
Expand
View Pricing
BAH369062008
SP Bel-Art
®
Sterileware
™
Sampling Scoop with Handle,
red polystyrene, nominal capacity 250 mL (8 oz), sterile, pack of 100 ea (individually wrapped)
Expand
View Pricing
BAH369041010
SP Bel-Art
®
Sterileware
™
Sampling Scoop with Handle,
white polystyrene, nominal capacity 125 mL (4 oz), sterile, pack of 100 ea (double bagged)
Expand
View Pricing
BAH369042004
SP Bel-Art
®
Sterileware
™
Sampling Scoop with Handle,
red polystyrene, nominal capacity 125 mL (4 oz), sterile, pack of 100 ea (individually wrapped)
Expand
View Pricing