Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
SP Bel-Art
®
Sterileware
™
Pharma Scoop
Products
Product Category
spatulas/scoops (14)
Manufacturer
Height (mm)
Length (in)
Length (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
BAF369090500
SP Bel-Art
®
Sterileware
™
Pharma Scoop,
white polystyrene, nominal capacity 500 mL (17 oz), sterile, pack of 50 ea (individually wrapped)
Expand
View Pricing
BAF369090125
SP Bel-Art
®
Sterileware
™
Pharma Scoop,
white polystyrene, nominal capacity 125 mL (4 oz), sterile, pack of 100 ea (individually wrapped)
Expand
View Pricing
BAF369091000
SP Bel-Art
®
Sterileware
™
Pharma Scoop,
white polystyrene, nominal capacity 1000 mL (34 oz), sterile, pack of 25 ea (individually wrapped)
Expand
View Pricing
BAF369092500
SP Bel-Art
®
Sterileware
™
Pharma Scoop,
white polystyrene, nominal capacity 2500 mL (85 oz), sterile, pack of 15 ea (individually wrapped)
Expand
View Pricing
BAF369102500
SP Bel-Art
®
Sterileware
™
Pharma Scoop,
red polystyrene, nominal capacity 2500 mL (85 oz), sterile, pack of 15 ea (individually wrapped)
Expand
View Pricing
BAF369090030
SP Bel-Art
®
Sterileware
™
Pharma Scoop,
white polystyrene, nominal capacity 30.0 mL (1 oz), sterile, pack of 100 ea (individually wrapped)
Expand
View Pricing
BAF369100030
SP Bel-Art
®
Sterileware
™
Pharma Scoop,
red polystyrene, nominal capacity 30 mL (1 oz), sterile, pack of 100 ea (individually wrapped)
Expand
View Pricing
BAF369090060
SP Bel-Art
®
Sterileware
™
Pharma Scoop,
white polystyrene, nominal capacity 60 mL (2 oz), sterile, pack of 100 ea (individually wrapped)
Expand
View Pricing
BAF369100500
SP Bel-Art
®
Sterileware
™
Pharma Scoop,
red polystyrene, nominal capacity 500 mL (17 oz), sterile, pack of 50 ea (individually wrapped)
Expand
View Pricing
BAF369090250
SP Bel-Art
®
Sterileware
™
Pharma Scoop,
white polystyrene, nominal capacity 250 mL (8 oz), sterile, pack of 80 ea (individually wrapped)
Expand
View Pricing
BAF369100250
SP Bel-Art
®
Sterileware
™
Pharma Scoop,
red polystyrene, nominal capacity 250 mL (8 oz), sterile, pack of 80 ea (individually wrapped)
Expand
View Pricing
BAF369100060
SP Bel-Art
®
Sterileware
™
Pharma Scoop,
red polystyrene, nominal capacity 60 mL (2 oz), sterile, pack of 100 ea (individually wrapped)
Expand
View Pricing
BAF369101000
SP Bel-Art
®
Sterileware
™
Pharma Scoop,
red polystyrene, nominal capacity 1000 mL (34 oz), sterile, pack of 25 ea (individually wrapped)
Expand
View Pricing
BAF369100125
SP Bel-Art
®
Sterileware
™
Pharma Scoop,
red polystyrene, nominal capacity 125 mL (4 oz), sterile, pack of 100 ea (individually wrapped)
Expand
View Pricing