Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Soybean oil
Synonym(s):
Soya oil from
Glycine max
, Soybean oil
CAS Number:
8001-22-7
Products
Product Category
lipids (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Biological Source
Manufacturer
Physical Form
Agency
Format
Quality Segment
Functional Group
Application
Package Size
API Family
Reference Material Type
Special Grade
certified reference material (1)
pharmaceutical primary standard (1)
pharmaceutical secondary standard (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
S7381
Soybean oil,
dietary source of long-chain triglycerides and other lipids
Expand
View Pricing
1617500
Soybean oil,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
PHR2882
Soybean oil,
pharmaceutical secondary standard, certified reference material
Expand
View Pricing