Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sorenson
™
low binding standard tips
Products
Product Category
pipette tips (11)
Manufacturer
Color
Package Size
Sterility
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
Z719668
Sorenson
™
low binding standard tips,
maximum volume 1000 μL, pack of 1000 ea (Bag), non-sterile
Expand
View Pricing
Z719684
Sorenson
™
low binding standard tips,
maximum volume 1000 μL, pack of 1000 ea (Racked), sterile
Expand
View Pricing
Z719579
Sorenson
™
low binding standard tips,
maximum volume 200 μL, pack of 1000 ea (bag), non-sterile
Expand
View Pricing
Z719595
Sorenson
™
low binding standard tips,
maximum volume 200 μL, pkg of 96 tips CellRack, sterile
Expand
View Pricing
Z719625
Sorenson
™
low binding standard tips,
maximum volume 300 μL, case of 1000 ea (Bag), non-sterile
Expand
View Pricing
Z719581
Sorenson
™
low binding standard tips,
maximum volume 200 μL, pkg of 96 tips CellRack, non-sterile
Expand
View Pricing
Z719676
Sorenson
™
low binding standard tips,
maximum volume 1000 μL, non-sterile
Expand
View Pricing
Z719617
Sorenson
™
low binding standard tips,
maximum volume 200 μL, pkg of 96 QuickRack, non-sterile
Expand
View Pricing
Z719706
Sorenson
™
low binding standard tips,
pack of 8 ea (Empty racks), non-sterile
Expand
View Pricing
Z719633
Sorenson
™
low binding standard tips,
maximum volume 300 μL, rack of 96, non-sterile
Expand
View Pricing
Z719641
Sorenson
™
low binding standard tips,
maximum volume 300 μL, rack of 96, sterile
Expand
View Pricing