Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium tetraphenyl borate
Synonym(s):
Sodium tetraphenyl borate, Tetraphenylboron sodium
Linear Formula:
(C
6
H
5
)
4
BNa
CAS Number:
143-66-8
Molecular Weight:
342.22
EC Number:
205-605-5
Products
Brand
Sigma-Aldrich (2)
Supelco (1)
Formula Weight
Physical Form
Purity
Agency
Analyte Suitability
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Special Grade
ACS reagent (2)
Puriss (1)
Puriss P.A. (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
1.06669
Sodium tetraphenyl borate,
GR for analysis ACS,Reag. Ph Eur
Expand
View Pricing
T25402
Sodium tetraphenyl borate,
ACS reagent, ≥99.5%
Expand
View Pricing
72020
Sodium tetraphenyl borate,
puriss. p.a., ACS reagent, ≥99.5% (NT)
Expand
View Pricing