Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium oleate
Synonym(s):
cis
-9-Octadecenoic acid sodium salt, Oleic acid sodium salt
Linear Formula:
CH
3
(CH
2
)
7
CH=CH(CH
2
)
7
COONa
CAS Number:
143-19-1
Molecular Weight:
304.44
EC Number:
205-591-0
Products
Product Category
fatty acids (3)
lipids (3)
metabolites (1)
Brand
Sigma-Aldrich (3)
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
O7501
Sodium oleate,
≥99%
Expand
View Pricing
26125
Sodium oleate,
≥82% (fatty acids), powder
Expand
View Pricing
O3880
Sodium oleate,
≥95% (capillary GC)
Expand
View Pricing