Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium iodide
Synonym(s):
Sodium iodide
Linear Formula:
NaI
CAS Number:
7681-82-5
Molecular Weight:
149.89
EC Number:
231-679-3
Products
Product Category
salts (10)
Brand
SAFC (1)
Sigma-Aldrich (8)
Supelco (2)
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
ACS reagent (2)
Anhydrous (2)
Puriss (1)
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
383112
Sodium iodide,
ACS reagent, ≥99.5%
Expand
View Pricing
217638
Sodium iodide,
ReagentPlus
®
, ≥99%
Expand
View Pricing
409286
Sodium iodide,
99.999% trace metals basis
Expand
View Pricing
1.06523
Sodium iodide,
for analysis EMSURE
®
ACS,Reag. Ph Eur
Expand
View Pricing
746371
Sodium iodide,
anhydrous, free-flowing, Redi-Dri
™
, ACS reagent, ≥99.5%
Expand
View Pricing
03129
Sodium iodide,
puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. to the dried substance)
Expand
View Pricing
1.06520
Sodium iodide,
EMPROVE
®
ESSENTIAL, Ph. Eur., BP, JP, USP
Expand
View Pricing
229911
Sodium iodide,
≥99.99% trace metals basis
Expand
View Pricing
1.06519
Sodium iodide,
99.99 Suprapur
®
Expand
View Pricing
793558
Sodium iodide,
anhydrous, free-flowing, Redi-Dri
™
,
ReagentPlus
®
, ≥99%
Expand
View Pricing
439681
Sodium iodide,
AnhydroBeads
™
, −10 mesh, 99.999% trace metals basis
Expand
View Pricing