Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium hypochlorite solution
Synonym(s):
Sodium hypochlorite solution
Linear Formula:
NaClO
CAS Number:
7681-52-9
Molecular Weight:
74.44
EC Number:
231-668-3
Products
Product Category
salts (2)
detergents (1)
Brand
Sigma-Aldrich (3)
Sustainability
Boiling Point (°C)
Feature
Formula Weight
pH Value
Physical Form
Product Line
Agency
Chemical Composition
Quality Segment
Reagent Type
Special Grade
Purum (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
425044
Sodium hypochlorite solution,
reagent grade, available chlorine 10-15 %
Expand
View Pricing
1.05614
Sodium hypochlorite solution,
(6-14% active chlorine) EMPLURA
®
Expand
View Pricing
13440
Sodium hypochlorite solution,
6-14% active chlorine basis
Expand
View Pricing
71696
Sodium hypochlorite solution,
purum, ~10% (RT)
Expand
View Pricing