Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium hydrosulfite
Synonym(s):
Sodium hypodisulfite
Linear Formula:
NaO
2
SSO
2
Na
CAS Number:
7775-14-6
Molecular Weight:
174.11
EC Number:
231-890-0
Products
Product Category
detergents (1)
salts (1)
Brand
Sigma-Aldrich (2)
Feature
Formula Weight
Physical Form
Purity
Quality Segment
Reagent Type
Special Grade
Technical (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
157953
Sodium hydrosulfite,
technical grade
Expand
View Pricing
71699
Sodium hydrosulfite,
≥82% (RT)
Expand
View Pricing