Skip to Content
Merck
Sodium D-pantoate Pharmaceutical Secondary Standard; Certified Reference Material

Sodium D-pantoate

Sodium D-pantoate Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
(R)-Pantoic acid sodium salt, (2R)-2,4-Dihydroxy-3,3-dimethylbutanoic acid monosodium salt, (R)-2,4-Dihydroxy-3,3-dimethylbutyric acid sodium salt, Sodium (R)-pantoate, Sodium D-pantoate
Empirical Formula (Hill Notation):
C6H11NaO4
CAS Number:
Molecular Weight:
170.14

Products

Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Optical Isomer
API Family
Reference Material Type
Special Grade