Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium carbonate decahydrate
Synonym(s):
Sal soda, Soda, Sodium carbonate decahydrate
Linear Formula:
Na
2
CO
3
· 10H
2
O
CAS Number:
6132-02-1
Molecular Weight:
286.14
EC Number:
207-838-8
Products
Product Category
salts (2)
inorganic bases (1)
Brand
Sigma-Aldrich (2)
Supelco (1)
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Special Grade
Puriss (1)
Puriss P.A. (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
71360
Sodium carbonate decahydrate,
puriss. p.a., ≥99.0% (T)
Expand
View Pricing
1.06391
Sodium carbonate decahydrate,
for analysis EMSURE
®
ISO,Reag. Ph Eur
Expand
View Pricing
577782
Sodium carbonate decahydrate,
99.999% trace metals basis
Expand
View Pricing