Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium bitartrate monohydrate
Synonym(s):
L-(+)-Tartaric acid monosodium salt, Sodium hydrogen tartrate monohydrate
Empirical Formula (Hill Notation):
C
4
H
5
NaO
6
· H
2
O
CAS Number:
6131-98-2
Molecular Weight:
190.08
EC Number:
208-400-9
Products
Product Category
buffers (1)
Brand
Sigma-Aldrich (1)
Formula Weight
pH Value
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Optical Isomer
Special Grade
Purum (1)
Purum P.A. (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
71679
Sodium bitartrate monohydrate,
BioUltra, ≥99.0% (T)
Expand
View Pricing
71680
Sodium bitartrate monohydrate,
purum p.a., ≥98% (T)
Expand
View Pricing