Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium bisulfite
Synonym(s):
Sodium hydrogensulfite
Empirical Formula (Hill Notation):
NaHSO
3
CAS Number:
7631-90-5
Molecular Weight:
104.06
EC Number:
231-548-0
Products
Product Category
salts (3)
Brand
Sigma-Aldrich (3)
Formula Weight
Physical Form
Product Line
Quality Segment
Special Grade
ACS reagent (2)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
243973
Sodium bisulfite,
ACS reagent
Expand
View Pricing
799394
Sodium bisulfite,
anhydrous, free-flowing, Redi-Dri
™
, ACS reagent
Expand
View Pricing
28-1970
Sodium bisulfite,
JIS special grade, 64.0-67.4% SO
2
basis
Expand
View Pricing