Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium
Synonym(s):
Sodium
Empirical Formula (Hill Notation):
Na
CAS Number:
7440-23-5
Molecular Weight:
22.99
EC Number:
231-132-9
Products
Product Category
metals (4)
Brand
Sigma-Aldrich (6)
Supelco (1)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reagent Type
Special Grade
ACS reagent (1)
Purum (1)
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
483745
Sodium,
cubes, contains mineral oil, 99.9% trace metals basis
Expand
View Pricing
71172
Sodium,
in kerosene, pieces (large), ≥99.8% (sodium basis)
Expand
View Pricing
262714
Sodium,
99.95% trace metals basis, ingot
Expand
View Pricing
8.22284
Sodium,
rods (protective liquid: paraffin oil) for synthesis
Expand
View Pricing
244686
Sodium,
25-35 wt % dispersion in paraffin
Expand
View Pricing
1.06260
Sodium,
rod diameter 2.5 cm (protective liquid: paraffin oil)
Expand
View Pricing
282065
Sodium,
ACS reagent, dry
Expand
View Pricing