Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Snap Ring polypropylene autosampler microvial
Products
Product Category
vials (5)
Brand
Supelco (5)
ID (mm)
OD (mm)
Volume (ml)
Closure Type
Color
Vial Bottle Material
Vial Bottle Color
Special Grade
Natural (4)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
27422
Snap Ring polypropylene autosampler microvial,
volume 0.5 mL, natural polypropylene vial, O.D. × H × I.D. 12 mm × 32 mm × 6 mm, Snap Ring, pkg of 100 ea
Expand
View Pricing
27423
Snap Ring polypropylene autosampler microvial,
volume 0.1 mL, natural polypropylene vial, O.D. × H × I.D. 12 mm × 32 mm × 6 mm, Snap Ring, pkg of 100 ea
Expand
View Pricing
27253
Snap Ring polypropylene autosampler microvial,
volume 0.75 mL, natural polypropylene vial, Snap top, pkg of 100 ea
Expand
View Pricing
27254
Snap Ring polypropylene autosampler microvial,
volume 0.75 mL, natural polypropylene vial, pkg of 1000 ea
Expand
View Pricing
SU860114
Snap Ring polypropylene autosampler microvial,
volume 0.3 mL
Expand
View Pricing