Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
SMCC Conjugation Kit for Amine Cd-based Core/Shell Quantum Dots
Products
Product Category
kits (9)
Brand
Sigma-Aldrich (9)
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
OCNQAK665
SMCC Conjugation Kit for Amine Cd-based Core/Shell Quantum Dots,
fluorescence λ
em
665 nm, for 4 conjugation
Expand
View Pricing
OCNQAK580
SMCC Conjugation Kit for Amine Cd-based Core/Shell Quantum Dots,
fluorescence λ
em
580 nm, for 4 conjugation
Expand
View Pricing
OCNQAK600
SMCC Conjugation Kit for Amine Cd-based Core/Shell Quantum Dots,
fluorescence λ
em
600 nm, for 4 conjugation
Expand
View Pricing
OCNQAK525
SMCC Conjugation Kit for Amine Cd-based Core/Shell Quantum Dots,
fluorescence λ
em
525 nm, for 4 conjugation
Expand
View Pricing
OCNQAK620
SMCC Conjugation Kit for Amine Cd-based Core/Shell Quantum Dots,
fluorescence λ
em
620 nm, for 4 conjugation
Expand
View Pricing
OCNQAK450
SMCC Conjugation Kit for Amine Cd-based Core/Shell Quantum Dots,
fluorescence λ
em
450 nm, for 4 conjugation
Expand
View Pricing
OCNQAK540
SMCC Conjugation Kit for Amine Cd-based Core/Shell Quantum Dots,
fluorescence λ
em
540 nm, for 4 conjugation
Expand
View Pricing
OCNQAK645
SMCC Conjugation Kit for Amine Cd-based Core/Shell Quantum Dots,
fluorescence λ
em
645 nm, for 4 conjugation
Expand
View Pricing
OCNQAK490
SMCC Conjugation Kit for Amine Cd-based Core/Shell Quantum Dots,
fluorescence λ
em
490 nm, for 4 conjugation
Expand
View Pricing