Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Silver
EC Number:
200-578-6
Products
Brand
Sigma-Aldrich (13)
Color
Physical Form
Quality Segment
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
807508
Silver,
Nanowires, diam. × L 50 nm (±10 nm) × 40 μm (±5 μm), 5 mg/mL (in IPA)
Expand
View Pricing
807591
Silver,
Nanowires, diam. × L 60 nm (±10 nm) × 40 μm (±5 μm), 5 mg/mL (in water)
Expand
View Pricing
806714
Silver,
Nanowires, diam. × L 20 nm (±2 nm) × 12 μm (±2 μm), 5 mg/mL (in IPA)
Expand
View Pricing
807826
Silver,
Nanowires, diam. × L 70 nm (±10 nm) × 40 μm (±5 μm), 5 mg/mL (in water)
Expand
View Pricing
806609
Silver,
nanowires, diam. × L 20 nm (±2 nm) × 12 μm (±2 μm), 5 mg/mL (in water)
Expand
View Pricing
807702
Silver,
Nanowires, diam. × L 60 nm (±10 nm) × 40 μm (±5 μm), 5 mg/mL (in IPA)
Expand
View Pricing
808644
Silver,
Nanowires, diam. × L 60±5 nm × 40±5 μm, 5 mg/mL in ETH
Expand
View Pricing
806722
Silver,
Nanowires, diam. × L 25 nm (±3 nm) × 15 μm (±2 μm), 5 mg/mL (in water)
Expand
View Pricing
807923
Silver,
Nanowires, diam. × L 70 nm (±10 nm) × 40 μm (±5 μm), 5 mg/mL (in ETH)
Expand
View Pricing
806935
Silver,
Nanowires, diam. × L 30 nm (±5 nm) × 20 μm (±3 μm), 5 mg/mL (in water)
Expand
View Pricing
806617
Silver,
Nanowires, diam. × L 20 nm (±2 nm) × 12 μm (±2 μm), 5 mg/mL (in ETH)
Expand
View Pricing
808032
Silver,
Nanowires, diam. × L 70 nm (±10 nm) × 40 μm (±5 μm), 5 mg/mL (in IPA)
Expand
View Pricing
807168
Silver,
Nanowires, diam. × L 35 nm (±5 nm) × 25 μm (±5 μm), 5 mg/mL (in ETH)
Expand
View Pricing