Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Silicon carbide
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Particle Size (µm)
Filter Products
Showing 1-20 of 28
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GF09680160
Silicon carbide,
powder, mean particle size 0.1-1 micron, weight 200 g, purity 99%
Expand
View Pricing
GF82027470
Silicon carbide,
sheet, thickness 4 mm, size 50 × 50 mm, condition direct sintered
Expand
View Pricing
GF10020339
Silicon carbide,
powder, mean particle size 0.1-1 micron, weight 500 g, purity 99%
Expand
View Pricing
GF30991293
Silicon carbide,
powder, mean particle size 17 micron, weight 50 g, green
Expand
View Pricing
GF06217006
Silicon carbide,
powder, max. particle size 0.05 micron, weight 100 g, purity 99%
Expand
View Pricing
GF48263014
Silicon carbide,
powder, mean particle size 0.1-1 micron, weight 50 g, purity 99%
Expand
View Pricing
GF32877445
Silicon carbide,
powder, mean particle size 0.1-1 micron, weight 100 g, purity 99%
Expand
View Pricing
GF67785856
Silicon carbide,
powder, 75 max. part. size (micron), weight 50 g, purity 98.7%
Expand
View Pricing
GF40471455
Silicon carbide,
powder, mean particle size 0.1-1 micron, weight 500 g, purity 99%
Expand
View Pricing
GF49923771
Silicon carbide,
powder, mean particle size 0.1-1 micron, weight 200 g, purity 99%
Expand
View Pricing
GF62680822
Silicon carbide,
monofilament, 0.1 mm diameter, length 1 m, core tungsten 0.01mm diameter
Expand
View Pricing
GF47017650
Silicon carbide,
fiber, tex number 209, length 10 m, filament diameter 0.015mm
Expand
View Pricing
GF02286389
Silicon carbide,
fiber, tex number 209, length 100 m, filament diameter 0.015mm
Expand
View Pricing
GF03897490
Silicon carbide,
monofilament, 0.1 mm diameter, length 1 m, core tungsten 0.01mm diameter
Expand
View Pricing
GF40770435
Silicon carbide,
fiber, tex number 209, length 1 m, filament diameter 0.015mm
Expand
View Pricing
GF08399391
Silicon carbide,
powder, mean particle size 0.1-1 micron, weight 100 g, purity 99%
Expand
View Pricing
GF37976147
Silicon carbide,
fiber, tex number 209, length 5 m, filament diameter 0.015mm
Expand
View Pricing
GF19379920
Silicon carbide,
powder, max. particle size 75 micron, weight 200 g, purity 98.7%
Expand
View Pricing
GF50814873
Silicon carbide,
fiber, tex number 209, length 50 m, filament diameter 0.015mm
Expand
View Pricing
GF22490423
Silicon carbide,
powder, max. particle size 75 micron, weight 500 g, purity 98.7%
Expand
View Pricing
Page 1 of 2
Page 1 of 2