Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Serrated natural rubber septa
Products
Product Category
septa (20)
Brand
Sigma-Aldrich (20)
OD (mm)
Color
Package Size
Special Grade
Natural (20)
Filter Products
Showing 1-20 of 20
Compare
Product Number
Description
Pricing
Z512141
Serrated natural rubber septa,
white rubber, bottom O.D. 23 mm, fits neck I.D., 20.5 mm, ST/NS: 24/40
Expand
View Pricing
Z512117
Serrated natural rubber septa,
red rubber, bottom O.D. 14.5 mm, fits neck I.D., 12.5 mm, ST/NS: 14/20
Expand
View Pricing
Z512133
Serrated natural rubber septa,
red rubber, bottom O.D. 23 mm, fits neck I.D., 20.5 mm, ST/NS: 24/40
Expand
View Pricing
Z512125
Serrated natural rubber septa,
white rubber, bottom O.D. 14.5 mm, fits neck I.D., 12.5 mm, ST/NS: 14/20
Expand
View Pricing
Z512222
Serrated natural rubber septa,
red rubber, bottom O.D. 28 mm, fits neck I.D., 25.5 mm, ST/NS: 29/42
Expand
View Pricing
Z512214
Serrated natural rubber septa,
white rubber, bottom O.D. 19.5 mm, fits neck I.D., 17.5 mm, ST/NS: 19/22
Expand
View Pricing
Z512206
Serrated natural rubber septa,
red rubber, bottom O.D. 19.5 mm, fits neck I.D., 17.5 mm, ST/NS: 19/22
Expand
View Pricing
Z512176
Serrated natural rubber septa,
white rubber, bottom O.D. 10 mm, fits neck I.D., 8.0 mm, ST/NS: 10/30
Expand
View Pricing
Z512168
Serrated natural rubber septa,
red rubber, bottom O.D. 10 mm, fits neck I.D., 8.0 mm
Expand
View Pricing
Z512230
Serrated natural rubber septa,
white rubber, bottom O.D. 28 mm, fits neck I.D., 25.5 mm, ST/NS: 29/42
Expand
View Pricing
Z512184
Serrated natural rubber septa,
red rubber, bottom O.D. 18 mm, fits neck I.D., 16.0 mm, Sure/Seal
Expand
View Pricing
Z512613
Serrated natural rubber septa,
white rubber, bottom O.D. 26.5 mm, fits neck I.D., 24.0 mm
Expand
View Pricing
Z512192
Serrated natural rubber septa,
white rubber, bottom O.D. 18 mm, fits neck I.D., 16.0 mm, Sure/Seal
Expand
View Pricing
Z512540
Serrated natural rubber septa,
red rubber, bottom O.D. 26.5 mm, fits neck I.D., 24.0 mm
Expand
View Pricing
Z512621
Serrated natural rubber septa,
blue rubber, bottom O.D. 8.5 mm, fits neck I.D., 6.5 mm
Expand
View Pricing
Z512656
Serrated natural rubber septa,
blue rubber, bottom O.D. 11.5 mm, fits neck I.D., 9.5 mm
Expand
View Pricing
Z512737
Serrated natural rubber septa,
blue rubber, bottom O.D. 25 mm, fits neck I.D., 22.0 mm
Expand
View Pricing
Z512524
Serrated natural rubber septa,
red rubber, bottom O.D. 21 mm, fits neck I.D., 19.0 mm
Expand
View Pricing
Z512710
Serrated natural rubber septa,
blue rubber, bottom O.D. 21 mm, fits neck I.D., 19.0 mm
Expand
View Pricing
Z512699
Serrated natural rubber septa,
blue rubber, bottom O.D. 18 mm, fits neck I.D., 16.0 mm, Sure/Seal
Expand
View Pricing