Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Septa, tan PTFE/silicone
Products
Product Category
septa (9)
Brand
Supelco (9)
Diameter (mm)
OD (mm)
Color
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
27188-U
Septa, tan PTFE/silicone,
white tan PTFE/silicone, diam. × thickness 22 mm × 0.100 in., for use with 20, 40 or 60 mL vial, pkg of 100 ea
Expand
View Pricing
27177
Septa, tan PTFE/silicone,
white tan PTFE/silicone, diam. × total thickness × PTFE thickness 18 mm × 0.060 in. × 5 mil, for use with 22 mL vial, pkg of 100 ea
Expand
View Pricing
23193-U
Septa, tan PTFE/silicone,
white tan PTFE/silicone, diam. × thickness 22 mm × 0.125 in., for use with 20, 40 or 60 mL vial, pkg of 100 ea
Expand
View Pricing
27155
Septa, tan PTFE/silicone,
white tan PTFE/silicone, diam. × thickness 13 mm × 0.060 in., for use with 7 mL vial, pkg of 100 ea
Expand
View Pricing
27144
Septa, tan PTFE/silicone,
white tan PTFE/silicone, diam. × thickness 11 mm × 0.060 in., for use with 4 mL vial, pkg of 100 ea
Expand
View Pricing
27166
Septa, tan PTFE/silicone,
white tan PTFE/silicone, diam. × thickness 16 mm × 0.090 in., for use with 15 mL vial, pkg of 100 ea
Expand
View Pricing
27380-U
Septa, tan PTFE/silicone,
tan PTFE/silicone, diam. × thickness 16 mm × 0.060 in., for use with 15 mL vial, pkg of 100 ea
Expand
View Pricing
27095-U
Septa, tan PTFE/silicone,
white tan PTFE/silicone, diam. × thickness 8 mm × 0.060 in., pkg of 100 ea
Expand
View Pricing
SU860007
Septa, tan PTFE/silicone,
tan PTFE/silicone, diam. × thickness 22 mm × 3.2 mm, for use with 20, 40, 60mL vial, pkg of 1000 ea
Expand
View Pricing