Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
SealMate
™
system for adhesive microplate-sealing films
Products
Product Category
sealing films and foils (8)
Color
Manufacturer
Length (m)
Color
Plate Material
Package Size
Sterility
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
Z725072
SealMate
™
system for adhesive microplate-sealing films,
AeraSeal
™
starter kit, includes SealMate
™
dispenser + 2 rolls of AeraSeal
™
film, sterile
Expand
View Pricing
Z724742
SealMate
™
system for adhesive microplate-sealing films,
SealPlate
®
starter kit, includes SealMate
™
dispenser + 2 rolls of SealPlate
®
film, non-sterile
Expand
View Pricing
Z724955
SealMate
™
system for adhesive microplate-sealing films,
AeraSeal
™
starter kit, includes SealMate
™
dispenser + 2 rolls of AeraSeal
™
film, non-sterile
Expand
View Pricing
Z726281
SealMate
™
system for adhesive microplate-sealing films,
AeraSeal
™
rolls, sterile
Expand
View Pricing
Z725625
SealMate
™
system for adhesive microplate-sealing films,
SealPlate
®
Rolls, non-sterile
Expand
View Pricing
Z726079
SealMate
™
system for adhesive microplate-sealing films,
AeraSeal
™
rolls, non-sterile
Expand
View Pricing
Z724858
SealMate
™
system for adhesive microplate-sealing films,
SealPlate
®
starter kit, includes SealMate
™
dispenser + 2 rolls of SealPlate
®
film, sterile
Expand
View Pricing
Z725854
SealMate
™
system for adhesive microplate-sealing films,
SealPlate
®
rolls, sterile
Expand
View Pricing