
S-Desoxybrevetoxin B2-(carboxyethyl-13C3,42-d,15N) solution

Empirical Formula (Hill Notation):
13C3C50DH7815NO16S
Molecular Weight:
1023.23
Products
Product Category
Brand
Formula Weight
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing