Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Screw cap tubes and caps with O-rings
Products
Product Category
tubes (5)
Color
Manufacturer
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
AXYSCT150C
Screw cap tubes and caps with O-rings,
size 1.5 mL, clear, pkg of 8x500caps/cs
Expand
View Pricing
AXYSCT200C
Screw cap tubes and caps with O-rings,
size 2.0 mL, clear, pkg of 8x500caps/cs
Expand
View Pricing
AXYSCT150Y
Screw cap tubes and caps with O-rings,
size 1.5 mL, yellow, pkg of 8x500caps/cs
Expand
View Pricing
AXYSCT200B
Screw cap tubes and caps with O-rings,
size 2.0 mL, blue, pkg of 8x500caps/cs
Expand
View Pricing
AXYSCT200W
Screw cap tubes and caps with O-rings,
size 2.0 mL, white, pkg of 8x500caps/cs
Expand
View Pricing