Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Screw cap tube
Products
Product Category
tubes (13)
Color
Manufacturer
Package Size
Sterility
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
AXYSCT150CS
Screw cap tube,
size 1.5 mL, clear, pkg of 8x500caps/cs, sterile
Expand
View Pricing
AXYSCT200SSCS
Screw cap tube,
self-standing, size 2.0 mL, clear, pkg of 8x500caps/cs, sterile
Expand
View Pricing
AXYSCT150AS
Screw cap tube,
size 1.5 mL, assorted colors, pkg of 8x500caps/cs, sterile
Expand
View Pricing
AXYSCT150SSCS
Screw cap tube,
self-standing, size 1.5 mL, clear, pkg of 8x500caps/cs, sterile
Expand
View Pricing
AXYST45S
Screw cap tube,
capacity 0.5 mL, clear, sterile
Expand
View Pricing
AXYSCT200CS
Screw cap tube,
size 2.0 mL, clear, pkg of 8x500caps/cs, sterile
Expand
View Pricing
AXYSCT050CS
Screw cap tube,
size 0.5 mL, clear, pkg of 8x500caps/cs, sterile
Expand
View Pricing
AXYSCT050SSCS
Screw cap tube,
self-standing, size 0.5 mL, clear, pkg of 8x500caps/cs, sterile
Expand
View Pricing
AXYSCT150SSAS
Screw cap tube,
self-standing, size 1.5 mL, assorted colors, pkg of 8x500caps/cs, sterile
Expand
View Pricing
AXYSCT200SSAS
Screw cap tube,
self-standing, size 2.0 mL, assorted colors, pkg of 8x500caps/cs, sterile
Expand
View Pricing
AXYSCT050AS
Screw cap tube,
size 0.5 mL, assorted colors, pkg of 8x500caps/cs, sterile
Expand
View Pricing
AXYSCT200AS
Screw cap tube,
size 2.0 mL, assorted colors, pkg of 8x500caps/cs, sterile
Expand
View Pricing
AXYSCT050SSAS
Screw cap tube,
self-standing, size 0.5 mL, assorted colors, pkg of 8x500caps/cs, sterile
Expand
View Pricing