Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Scienceware
®
Secador
®
desiccator cabinet
Products
Product Category
desiccators (9)
Brand
Sigma-Aldrich (4)
Color
Manufacturer
Package Size
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
Z553808
Scienceware
®
Secador
®
desiccator cabinet,
model 3.0, vertical profile, clear
Expand
View Pricing
Z553220
Scienceware
®
Secador
®
desiccator cabinet,
model 4.0, vertical profile, clear
Expand
View Pricing
Z553255
Scienceware
®
Secador
®
desiccator cabinet,
model 1.0, horizontal profile, clear
Expand
View Pricing
Z553786
Scienceware
®
Secador
®
desiccator cabinet,
model 2.0, horizontal profile, clear
Expand
View Pricing
Z566756
Scienceware
®
Secador
®
desiccator cabinet,
model 1.0, horizontal profile, amber
Expand
View Pricing
Z566772
Scienceware
®
Secador
®
desiccator cabinet,
model 3.0, vertical profile, amber
Expand
View Pricing
Z566780
Scienceware
®
Secador
®
desiccator cabinet,
model 4.0, vertical profile, amber
Expand
View Pricing
Z566764
Scienceware
®
Secador
®
desiccator cabinet,
model 2.0, horizontal profile, amber
Expand
View Pricing
Z553247
Scienceware
®
Secador
®
desiccator cabinet,
model 4.0, horizontal profile, clear
Expand
View Pricing