Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Scienceware
®
scraper
Products
Product Category
spatulas/scoops (6)
Brand
Sigma-Aldrich (3)
Manufacturer
Length (in)
Color
Package Size
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
Z500062
Scienceware
®
scraper,
white low-density polyethylene, handle L 10 in.
Expand
View Pricing
Z500038
Scienceware
®
scraper,
handle L 10 in., red high-density polyethylene
Expand
View Pricing
Z500046
Scienceware
®
scraper,
handle L 36 in., red high-density polyethylene
Expand
View Pricing
Z500089
Scienceware
®
scraper,
white low-density polyethylene, handle L 72 in.
Expand
View Pricing
Z500070
Scienceware
®
scraper,
white low-density polyethylene, handle L 36 in.
Expand
View Pricing
Z500054
Scienceware
®
scraper,
handle L 72 in., red high-density polyethylene
Expand
View Pricing