Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Scienceware
®
POXYGRID
®
test-tube rack
Products
Product Category
racks (13)
Brand
Sigma-Aldrich (6)
Manufacturer
Height (mm)
Length (cm)
Length (mm)
Color
Package Size
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
Z135801
Scienceware
®
POXYGRID
®
test-tube rack,
Holds 24 x 15-16 mm tubes
Expand
View Pricing
Z135798
Scienceware
®
POXYGRID
®
test-tube rack,
Holds 36 x 10-13 mm tubes
Expand
View Pricing
BAF187630160
Scienceware
®
POXYGRID
®
test-tube rack,
Holds 60 x 18-20 mm tubes, blue
Expand
View Pricing
Z178012
Scienceware
®
POXYGRID
®
test-tube rack,
Holds 108 x 15-16 mm tubes
Expand
View Pricing
BAF187560160
Scienceware
®
POXYGRID
®
test-tube rack,
Holds 60 x 13-16 mm tubes
Expand
View Pricing
Z178020
Scienceware
®
POXYGRID
®
test-tube rack,
Holds 40 x 22-25 mm tubes
Expand
View Pricing
Z177970
Scienceware
®
POXYGRID
®
test-tube rack,
Holds 72 x 10-13 mm tubes
Expand
View Pricing
BAF187521150
Scienceware
®
POXYGRID
®
test-tube rack,
Holds 150 x 10-13 mm tubes, blue
Expand
View Pricing
BAF187740160
Scienceware
®
POXYGRID
®
test-tube rack,
Holds 60 x 22-25 mm tubes, blue
Expand
View Pricing
Z177997
Scienceware
®
POXYGRID
®
test-tube rack,
Holds 48 x 15-16 mm tubes
Expand
View Pricing
Z135828
Scienceware
®
POXYGRID
®
test-tube rack,
Holds 20 x 18-20 mm tubes
Expand
View Pricing
BAF187510160
Scienceware
®
POXYGRID
®
test-tube rack,
Holds 60 x 10-13 mm tubes, blue
Expand
View Pricing
BAF187601150
Scienceware
®
POXYGRID
®
test-tube rack,
Holds 60 x 15-16 mm tubes, blue
Expand
View Pricing