Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sartorius pipette tips
Products
Product Category
pipette tips (28)
Manufacturer
Volume Range (µL)
Color
Package Size
Sterility
Filter Products
Showing 1-20 of 28
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
T6948
Sartorius pipette tips,
volume range 100-5000 μL, standard, bulk packed, non-sterile
Expand
View Pricing
Z709999
Sartorius pipette tips,
volume range 10-1000 μL, wide-bore, racked, sterile
Expand
View Pricing
Z741649
Sartorius pipette tips,
volume range 100-5000 μL, Standard, rack, sterile
Expand
View Pricing
Z709964
Sartorius pipette tips,
volume range 10-1000 μL, standard, racked, sterile
Expand
View Pricing
Z709972
Sartorius pipette tips,
volume range 10-1000 μL, standard, refill, non-sterile
Expand
View Pricing
Z757705
Sartorius pipette tips,
volume range 0.5-200 μL, standard, refill, non-sterile
Expand
View Pricing
Z709875
Sartorius pipette tips,
volume range 0.2-10 μL, standard, refill, non-sterile
Expand
View Pricing
Z757675
Sartorius pipette tips,
volume range 0.5-200 μL, standard, racked, non-sterile
Expand
View Pricing
Z741650
Sartorius pipette tips,
volume range 100-5000 μL, Standard, bulk, non-sterile
Expand
View Pricing
Z709921
Sartorius pipette tips,
volume range 5-350 μL, standard, racked, sterile
Expand
View Pricing
Z741651
Sartorius pipette tips,
volume range 500-10000 μL, Standard, bulk, non-sterile
Expand
View Pricing
Z709948
Sartorius pipette tips,
volume range 5-350 μL, standard, refill, non-sterile
Expand
View Pricing
Z757748
Sartorius pipette tips,
volume range 10-1000 μL, standard, refill, sterile
Expand
View Pricing
Z709980
Sartorius pipette tips,
volume range 10-1000 μL, wide-bore, racked, non-sterile
Expand
View Pricing
Z709956
Sartorius pipette tips,
volume range 10-1000 μL, standard, racked, non-sterile
Expand
View Pricing
Z757683
Sartorius pipette tips,
volume range 0.5-200 μL, standard, racked, sterile
Expand
View Pricing
Z709859
Sartorius pipette tips,
volume range 0.1-10 μL, standard, racked, non-sterile
Expand
View Pricing
Z741659
Sartorius pipette tips,
volume range 0.1-10 μL, Standard, non-sterile, bulk, pack of 1000 ea
Expand
View Pricing
Z741648
Sartorius pipette tips,
volume range 100-5000 μL, Standard, rack, non-sterile
Expand
View Pricing
Z709913
Sartorius pipette tips,
volume range 5-350 μL, standard, racked, non-sterile
Expand
View Pricing
Page 1 of 2
Page 1 of 2