Skip to Content
Merck
Saquinavir mesylate ≥98% (HPLC), powder

Saquinavir mesylate

Saquinavir mesylate ≥98% (HPLC), powder
Synonym(s):
(2S)-N1[(1S,2R)-3-[(3S,4aS,8aS)-3-[[(1,1-Dimethylethyl)amino]carbonyl]octahydro-2(1H)-isoquinolinyl]-2-hydroxy-1-(phenylmethyl)propyl]-2-[(2-quinolinylcarbonyl)amino]butanediamide methanesulfonate, Ro-31-8959
Empirical Formula (Hill Notation):
C38H50N6O5·CH4O3S
CAS Number:
Molecular Weight:
766.95

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2